Assay Details
| Gene Symbol: chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212824.2 | NP_997989.2 | 2 - 3 | 294 | 101 | |
| XM_005168522.4 | XP_005168579.1 | 2 - 3 | 492 | 101 | ||
| GenBank mRNA | AB097217.1 | - | 2 - 3 | 287 | 101 | |
| BC124140.1 | - | 2 - 3 | 294 | 101 | ||
| BC164169.1 | - | 2 - 3 | 238 | 101 | ||
More Information
Set Membership: |
|

chst11
Genomic Map