Assay Details
| Gene Symbol: sp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212863.2 | NP_998028.1 | 3 - 4 | 1083 | 52 | |
| GenBank mRNA | AY380818.1 | - | 1 - 2 | 820 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sp7| Gene Symbol: sp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212863.2 | NP_998028.1 | 3 - 4 | 1083 | 52 | |
| GenBank mRNA | AY380818.1 | - | 1 - 2 | 820 | 52 | |
Set Membership: |
|