Assay Details
| Gene Symbol: rbm24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212865.1 | NP_998030.1 | 1 - 2 | 174 | 111 | |
| GenBank mRNA | AB080735.1 | - | 1 - 2 | 174 | 111 | |
| BC163027.1 | - | 1 - 2 | 261 | 111 | ||
| BC163034.1 | - | 1 - 2 | 261 | 111 | ||
More Information
Set Membership: |
|

rbm24a
Genomic Map