Assay Details
| Gene Symbol: wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212920.1 | NP_998085.1 | 3 - 4 | 1002 | 69 | |
| GenBank mRNA | BC067729.1 | - | 3 - 4 | 1002 | 69 | |
More Information
Set Membership: |
|

wrnip1
Genomic Map