Assay Details
| Gene Symbol: fat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212967.2 | NP_998132.2 | 20 - 21 | 11629 | 73 | |
| XM_005168343.4 | XP_005168400.1 | 21 - 22 | 11743 | 73 | ||
| XM_005168345.4 | XP_005168402.1 | 21 - 22 | 11743 | 73 | ||
| XM_021469206.1 | XP_021324881.1 | 21 - 22 | 11743 | 73 | ||
| XM_021469232.1 | XP_021324907.1 | 21 - 22 | 11852 | 73 | ||
| XM_021469270.1 | XP_021324945.1 | 20 - 21 | 11674 | 73 | ||
| GenBank mRNA | AF469608.1 | - | 20 - 21 | 11627 | 73 | |
More Information
Set Membership: |
|

fat1a
Genomic Map