Assay Details
| Gene Symbol: rangap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212989.2 | NP_998154.2 | 12 - 13 | 1430 | 103 | |
| XM_005156154.4 | XP_005156211.1 | 12 - 13 | 1421 | 103 | ||
| GenBank mRNA | BC054654.1 | - | 12 - 13 | 1400 | 103 | |
| BC155267.1 | - | 12 - 13 | 1402 | 103 | ||
More Information
Set Membership: |
|

rangap1b
Genomic Map