Assay Details
| Gene Symbol: cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213080.2 | NP_998245.2 | 6 - 7 | 955 | 66 | |
| XM_009298475.3 | XP_009296750.1 | 6 - 7 | 945 | 66 | ||
| GenBank mRNA | BC054907.1 | - | 6 - 7 | 916 | 66 | |
More Information
Set Membership: |
|

cdc20
Genomic Map