Assay Details
| Gene Symbol: ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213129.2 | NP_998294.1 | 7 - 8 | 971 | 58 | |
| XM_005171827.2 | XP_005171884.1 | 8 - 9 | 1069 | 58 | ||
| GenBank mRNA | BC053275.1 | - | 7 - 8 | 973 | 58 | |
| BC152279.1 | - | 8 - 9 | 1017 | 58 | ||
More Information
Set Membership: |
|

ankra2
Genomic Map