Assay Details
| Gene Symbol: stap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213309.1 | NP_998474.1 | 3 - 4 | 436 | 72 | |
| XM_021478235.1 | XP_021333910.1 | 3 - 4 | 557 | 72 | ||
| GenBank mRNA | BC066612.1 | - | 3 - 4 | 436 | 72 | |
More Information
Set Membership: |
|

stap2b
Genomic Map