Assay Details
| Gene Symbol: eloa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213455.1 | NP_998620.1 | 350 | 62 | ||
| GenBank mRNA | BC044429.1 | - | 350 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eloa| Gene Symbol: eloa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213455.1 | NP_998620.1 | 350 | 62 | ||
| GenBank mRNA | BC044429.1 | - | 350 | 62 | ||
Set Membership: |
|