Assay Details
| Gene Symbol: chmp4bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213457.1 | NP_998622.1 | 1 - 2 | 265 | 93 | |
| GenBank mRNA | BC044191.1 | - | 1 - 2 | 265 | 93 | |
| BC071537.1 | - | 1 - 2 | 263 | 93 | ||
| BC165053.1 | - | 1 - 2 | 220 | 93 | ||
More Information
Set Membership: |
|

chmp4bb
Genomic Map