Assay Details
| Gene Symbol: aqp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213468.1 | NP_998633.1 | 2 - 3 | 301 | 75 | |
| GenBank mRNA | BC044188.1 | - | 2 - 3 | 301 | 75 | |
| BC164238.1 | - | 2 - 3 | 264 | 75 | ||
| EU341833.1 | - | 2 - 3 | 278 | 75 | ||
More Information
Set Membership: |
|

aqp3a
Genomic Map