Assay Details
| Gene Symbol: abcd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213482.1 | NP_998647.1 | 4 - 5 | 509 | 96 | |
| GenBank mRNA | BC044468.1 | - | 4 - 5 | 509 | 96 | |
More Information
Set Membership: |
|

abcd3a
Genomic Map