Assay Details
| Gene Symbol: abcd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213482.1 | NP_998647.1 | 6 - 7 | 666 | 80 | |
| XM_021470125.1 | XP_021325800.1 | 6 - 7 | 577 | 80 | ||
| GenBank mRNA | BC044468.1 | - | 6 - 7 | 666 | 80 | |
More Information
Set Membership: |
|

abcd3a
Genomic Map