Assay Details
| Gene Symbol: asmtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213511.1 | NP_998676.1 | 5 - 6 | 412 | 94 | |
| GenBank mRNA | BC055218.1 | - | 5 - 6 | 412 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

asmtl| Gene Symbol: asmtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213511.1 | NP_998676.1 | 5 - 6 | 412 | 94 | |
| GenBank mRNA | BC055218.1 | - | 5 - 6 | 412 | 94 | |
Set Membership: |
|