Assay Details
| Gene Symbol: emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213544.1 | NP_998709.1 | 8 - 9 | 613 | 66 | |
| GenBank mRNA | AY398368.1 | - | 8 - 9 | 613 | 66 | |
| BC071517.1 | - | 8 - 9 | 610 | 66 | ||
More Information
Set Membership: |
|

emc2
Genomic Map