Assay Details
| Gene Symbol: arhgef18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214818.1 | NP_999983.1 | 13 - 14 | 1792 | 74 | |
| XM_005171284.4 | XP_005171341.1 | 12 - 13 | 1835 | 74 | ||
| XM_009298637.3 | XP_009296912.1 | 14 - 15 | 1899 | 74 | ||
| XM_021479667.1 | XP_021335342.1 | 13 - 14 | 1872 | 74 | ||
| GenBank mRNA | BC070002.1 | - | 13 - 14 | 1792 | 74 | |
More Information
Set Membership: |
|

arhgef18a
Genomic Map