Assay Details
| Gene Symbol: arhgef18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214818.1 | NP_999983.1 | 16 - 17 | 2079 | 103 | |
| XM_005171284.4 | XP_005171341.1 | 15 - 16 | 2122 | 103 | ||
| XM_009298637.3 | XP_009296912.1 | 17 - 18 | 2186 | 103 | ||
| XM_021479667.1 | XP_021335342.1 | 16 - 17 | 2159 | 103 | ||
| GenBank mRNA | BC070002.1 | - | 16 - 17 | 2079 | 103 | |
More Information
Set Membership: |
|

arhgef18a
Genomic Map