Assay Details
| Gene Symbol: arhgef18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214818.1 | NP_999983.1 | 18 - 19 | 2571 | 151 | |
| XM_005171284.4 | XP_005171341.1 | 17 - 18 | 2614 | 151 | ||
| XM_009298637.3 | XP_009296912.1 | 19 - 20 | 2678 | 151 | ||
| XM_021479667.1 | XP_021335342.1 | 18 - 19 | 2651 | 151 | ||
| GenBank mRNA | BC070002.1 | - | 18 - 19 | 2571 | 151 | |
More Information
Set Membership: |
|

arhgef18a
Genomic Map