Assay Details
| Gene Symbol: rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001828.4 | NP_001001828.2 | 1 - 2 | 626 | 61 | |
| GenBank mRNA | AY609381.4 | - | 1 - 2 | 626 | 61 | |
| BC079515.1 | - | 1 - 2 | 173 | 61 | ||
More Information
Set Membership: |
|

rnf114
Genomic Map