Assay Details
| Gene Symbol: puraa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001846.1 | NP_001001846.1 | 1 | 355 | 75 | |
| GenBank mRNA | AY450553.1 | - | 1 | 355 | 75 | |
| BC081472.1 | - | 2 | 554 | 75 | ||
| BC151877.1 | - | 1 | 545 | 75 | ||
| BC164647.1 | - | 1 | 372 | 75 | ||
More Information
Set Membership: |
|

puraa
Genomic Map