Assay Details
| Gene Symbol: hdgfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002037.2 | NP_001002037.2 | 3 - 4 | 344 | 84 | |
| XM_009296311.3 | XP_009294586.1 | 3 - 4 | 429 | 84 | ||
| GenBank mRNA | AY728142.1 | - | 3 - 4 | 344 | 84 | |
| BC044157.1 | - | 3 - 4 | 417 | 84 | ||
| BC055551.1 | - | 3 - 4 | 410 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055551.1 transcripts.
Set Membership: |
|

hdgfl2
Genomic Map