Assay Details
| Gene Symbol: hdgfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002037.2 | NP_001002037.2 | 7 - 8 | 904 | 62 | |
| XM_009296311.3 | XP_009294586.1 | 7 - 8 | 986 | 62 | ||
| GenBank mRNA | AY728142.1 | - | 7 - 8 | 904 | 62 | |
| BC044157.1 | - | 7 - 8 | 977 | 62 | ||
More Information
Set Membership: |
|

hdgfl2
Genomic Map