Assay Details
| Gene Symbol: mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002095.1 | NP_001002095.1 | 8 - 9 | 775 | 89 | |
| XM_017354993.2 | XP_017210482.1 | 7 - 8 | 587 | 89 | ||
| XM_021474987.1 | XP_021330662.1 | 7 - 8 | 562 | 89 | ||
| XM_021474988.1 | XP_021330663.1 | 6 - 7 | 461 | 89 | ||
| GenBank mRNA | BC071526.1 | - | 8 - 9 | 775 | 89 | |
| JQ513868.1 | - | 8 - 9 | 676 | 89 | ||
| JQ513869.1 | - | 7 - 8 | 853 | 89 | ||
More Information
Set Membership: |
|

mapk11
Genomic Map