Assay Details
| Gene Symbol: mpdu1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002130.1 | NP_001002130.1 | 1 - 2 | 138 | 83 | |
| GenBank mRNA | BC071434.1 | - | 1 - 2 | 138 | 83 | |
More Information
Set Membership: |
|

mpdu1b
Genomic Map