Assay Details
| Gene Symbol: psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002131.1 | NP_001002131.1 | 3 - 4 | 343 | 85 | |
| GenBank mRNA | BC071432.1 | - | 3 - 4 | 343 | 85 | |
| JX971123.1 | - | 3 - 4 | 286 | 85 | ||
More Information
Set Membership: |
|

psmd8
Genomic Map