Assay Details
| Gene Symbol: sh3bgrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002197.1 | NP_001002197.1 | 2 - 3 | 379 | 145 | |
| GenBank mRNA | BC074037.1 | - | 2 - 3 | 379 | 145 | |
More Information
Set Membership: |
|

sh3bgrl2
Genomic Map