Assay Details
| Gene Symbol: sh3bgrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002197.1 | NP_001002197.1 | 3 - 4 | 442 | 97 | |
| GenBank mRNA | BC074037.1 | - | 3 - 4 | 442 | 97 | |
More Information
Set Membership: |
|

sh3bgrl2
Genomic Map