Assay Details
| Gene Symbol: acaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002207.1 | NP_001002207.1 | 3 - 4 | 315 | 77 | |
| GenBank mRNA | BC072706.1 | - | 3 - 4 | 315 | 77 | |
| BC165883.1 | - | 3 - 4 | 344 | 77 | ||
More Information
Set Membership: |
|

acaa1
Genomic Map