Assay Details
| Gene Symbol: kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002335.1 | NP_001002335.1 | 6 - 7 | 710 | 59 | |
| GenBank mRNA | BC075790.1 | - | 6 - 7 | 710 | 59 | |
More Information
Set Membership: |
|

kpna2
Genomic Map