Assay Details
| Gene Symbol: acy3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002347.1 | NP_001002347.1 | 5 - 6 | 807 | 128 | |
| XM_021470953.1 | XP_021326628.1 | 4 - 5 | 648 | 128 | ||
| GenBank mRNA | BC075914.1 | - | 5 - 6 | 807 | 128 | |
More Information
Set Membership: |
|

acy3.2
Genomic Map