Assay Details
| Gene Symbol: ppfibp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002362.2 | NP_001002362.2 | 2 - 3 | 178 | 75 | |
| NM_001326598.1 | NP_001313527.1 | 2 - 3 | 573 | 75 | ||
| GenBank mRNA | BC075891.1 | - | 2 - 3 | 178 | 75 | |
More Information
Set Membership: |
|

ppfibp2b
Genomic Map