Assay Details
| Gene Symbol: dnase1l1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002403.1 | NP_001002403.1 | 6 - 7 | 609 | 71 | |
| GenBank mRNA | BC076179.1 | - | 6 - 7 | 609 | 71 | |
More Information
Set Membership: |
|

dnase1l1l
Genomic Map