Assay Details
Gene Symbol: gnb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002437.1 | NP_001002437.1 | 4 - 5 | 501 | 82 | |
XM_021466676.1 | XP_021322351.1 | 4 - 5 | 425 | 82 | ||
GenBank mRNA | BC076127.1 | - | 4 - 5 | 501 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
