Assay Details
| Gene Symbol: syt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327829.1 | NP_001314758.1 | 4 - 5 | 507 | 62 | |
| XM_005164872.4 | XP_005164929.1 | 3 - 4 | 544 | 62 | ||
| XM_017354995.2 | XP_017210484.1 | 5 - 6 | 763 | 62 | ||
| XM_017354996.2 | XP_017210485.1 | 5 - 6 | 759 | 62 | ||
| GenBank mRNA | BC076355.1 | - | 3 - 4 | 515 | 62 | |
| BC164744.1 | - | 2 - 3 | 371 | 62 | ||
More Information
Set Membership: |
|

syt1a
Genomic Map