Assay Details
| Gene Symbol: syt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327829.1 | NP_001314758.1 | 5 - 6 | 632 | 110 | |
| XM_005164872.4 | XP_005164929.1 | 4 - 5 | 669 | 110 | ||
| XM_005164874.4 | XP_005164931.1 | 4 - 5 | 660 | 110 | ||
| XM_009300373.3 | XP_009298648.1 | 6 - 7 | 880 | 110 | ||
| XM_017354995.2 | XP_017210484.1 | 6 - 7 | 888 | 110 | ||
| XM_017354996.2 | XP_017210485.1 | 6 - 7 | 884 | 110 | ||
| GenBank mRNA | BC076355.1 | - | 4 - 5 | 640 | 110 | |
| BC164744.1 | - | 3 - 4 | 496 | 110 | ||
More Information
Set Membership: |
|

syt1a
Genomic Map