Assay Details
| Gene Symbol: syt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327829.1 | NP_001314758.1 | 6 - 7 | 802 | 105 | |
| XM_005164872.4 | XP_005164929.1 | 5 - 6 | 839 | 105 | ||
| XM_005164874.4 | XP_005164931.1 | 5 - 6 | 830 | 105 | ||
| XM_009300373.3 | XP_009298648.1 | 7 - 8 | 1050 | 105 | ||
| XM_017354995.2 | XP_017210484.1 | 7 - 8 | 1058 | 105 | ||
| XM_017354996.2 | XP_017210485.1 | 7 - 8 | 1054 | 105 | ||
| GenBank mRNA | BC076355.1 | - | 5 - 6 | 810 | 105 | |
| BC164744.1 | - | 4 - 5 | 666 | 105 | ||
More Information
Set Membership: |
|

syt1a
Genomic Map