Assay Details
| Gene Symbol: camk2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002542.1 | NP_001002542.1 | 14 - 15 | 1289 | 77 | |
| XM_005159836.4 | XP_005159893.1 | 14 - 15 | 1327 | 77 | ||
| XM_009306657.3 | XP_009304932.1 | 15 - 16 | 1393 | 77 | ||
| XM_009306661.3 | XP_009304936.1 | 15 - 16 | 1389 | 77 | ||
| XM_009306662.3 | XP_009304937.1 | 14 - 15 | 1332 | 77 | ||
| XM_009306663.3 | XP_009304938.1 | 14 - 15 | 1329 | 77 | ||
| XM_009306665.3 | XP_009304940.1 | 15 - 16 | 1391 | 77 | ||
| XM_009306667.3 | XP_009304942.1 | 15 - 16 | 1388 | 77 | ||
| XM_009306669.3 | XP_009304944.1 | 15 - 16 | 1391 | 77 | ||
| XM_017354018.2 | XP_017209507.1 | 15 - 16 | 1392 | 77 | ||
| XM_017354084.2 | XP_017209573.1 | 15 - 16 | 1387 | 77 | ||
| XM_021471362.1 | XP_021327037.1 | 15 - 16 | 1392 | 77 | ||
| XM_021471546.1 | XP_021327221.1 | 14 - 15 | 1331 | 77 | ||
| XM_021471587.1 | XP_021327262.1 | 15 - 16 | 1388 | 77 | ||
| GenBank mRNA | BC076266.1 | - | 14 - 15 | 1289 | 77 | |
| BC154768.1 | - | 14 - 15 | 1244 | 77 | ||
More Information
Set Membership: |
|

camk2d2
Genomic Map