Assay Details
| Gene Symbol: camk2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002542.1 | NP_001002542.1 | 16 - 17 | 1408 | 85 | |
| XM_005159836.4 | XP_005159893.1 | 16 - 17 | 1446 | 85 | ||
| XM_009306657.3 | XP_009304932.1 | 19 - 20 | 1635 | 85 | ||
| XM_009306661.3 | XP_009304936.1 | 19 - 20 | 1631 | 85 | ||
| XM_009306662.3 | XP_009304937.1 | 18 - 19 | 1574 | 85 | ||
| XM_009306663.3 | XP_009304938.1 | 18 - 19 | 1571 | 85 | ||
| XM_009306665.3 | XP_009304940.1 | 18 - 19 | 1531 | 85 | ||
| XM_009306667.3 | XP_009304942.1 | 18 - 19 | 1528 | 85 | ||
| XM_009306669.3 | XP_009304944.1 | 17 - 18 | 1510 | 85 | ||
| XM_017354018.2 | XP_017209507.1 | 19 - 20 | 1634 | 85 | ||
| XM_017354084.2 | XP_017209573.1 | 17 - 18 | 1506 | 85 | ||
| XM_021471362.1 | XP_021327037.1 | 18 - 19 | 1613 | 85 | ||
| XM_021471546.1 | XP_021327221.1 | 17 - 18 | 1471 | 85 | ||
| XM_021471587.1 | XP_021327262.1 | 17 - 18 | 1507 | 85 | ||
| GenBank mRNA | BC076266.1 | - | 16 - 17 | 1408 | 85 | |
| BC154768.1 | - | 16 - 17 | 1363 | 85 | ||
More Information
Set Membership: |
|

camk2d2
Genomic Map