Assay Details
| Gene Symbol: hid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002739.1 | NP_001002739.1 | 17 - 18 | 2254 | 86 | |
| GenBank mRNA | BC076413.1 | - | 17 - 18 | 2254 | 86 | |
More Information
Set Membership: |
|

hid1b
Genomic Map