Assay Details
| Gene Symbol: stx2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003561.1 | NP_001003561.1 | 5 - 6 | 421 | 112 | |
| GenBank mRNA | BC077121.1 | - | 5 - 6 | 421 | 112 | |
More Information
Set Membership: |
|

stx2a
Genomic Map