Assay Details
| Gene Symbol: myo6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004110.1 | NP_001004110.1 | 23 - 24 | 2430 | 78 | |
| XM_005158836.4 | XP_005158893.1 | 24 - 25 | 2684 | 78 | ||
| XM_009293188.3 | XP_009291463.1 | 23 - 24 | 2675 | 78 | ||
| XM_021467147.1 | XP_021322822.1 | 24 - 25 | 2688 | 78 | ||
| XM_021467148.1 | XP_021322823.1 | 23 - 24 | 2678 | 78 | ||
| GenBank mRNA | AY618701.1 | - | 23 - 24 | 2430 | 78 | |
More Information
Set Membership: |
|

myo6b
Genomic Map