Assay Details
| Gene Symbol: myo6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004110.1 | NP_001004110.1 | 29 - 30 | 3147 | 72 | |
| XM_021467147.1 | XP_021322822.1 | 30 - 31 | 3405 | 72 | ||
| XM_021467148.1 | XP_021322823.1 | 29 - 30 | 3395 | 72 | ||
| GenBank mRNA | AY618701.1 | - | 29 - 30 | 3147 | 72 | |
More Information
Set Membership: |
|

myo6b
Genomic Map