Assay Details
| Gene Symbol: sephs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004295.2 | NP_001004295.2 | 2 - 3 | 525 | 103 | |
| GenBank mRNA | AY221265.1 | - | 2 - 3 | 103 | ||
| BC081590.1 | - | 2 - 3 | 508 | 103 | ||
| CK146303.1 | - | 2 - 3 | 103 | |||
More Information
Set Membership: |
|

sephs3
Genomic Map