Assay Details
| Gene Symbol: sephs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004295.2 | NP_001004295.2 | 5 - 6 | 875 | 94 | |
| GenBank mRNA | AY221265.1 | - | 5 - 6 | 850 | 94 | |
| BC081590.1 | - | 5 - 6 | 858 | 94 | ||
More Information
Set Membership: |
|

sephs3
Genomic Map