Assay Details
| Gene Symbol: nudt4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004648.1 | NP_001004648.1 | 4 - 5 | 568 | 59 | |
| GenBank mRNA | BC080840.1 | - | 4 - 5 | 568 | 59 | |
More Information
Set Membership: |
|

nudt4b
Genomic Map