Assay Details
| Gene Symbol: lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004672.2 | NP_001004672.1 | 2 - 3 | 239 | 61 | |
| NM_001362514.1 | NP_001349443.1 | 2 - 3 | 279 | 61 | ||
| GenBank mRNA | AY584748.1 | - | 2 - 3 | 239 | 61 | |
| BC081422.1 | - | 2 - 3 | 257 | 61 | ||
| BC165911.1 | - | 2 - 3 | 195 | 61 | ||
More Information
Set Membership: |
|

lin7c
Genomic Map