Assay Details
| Gene Symbol: eloca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004673.1 | NP_001004673.1 | 4 - 5 | 391 | 71 | |
| XM_005162638.3 | XP_005162695.1 | 3 - 4 | 278 | 71 | ||
| XM_005162640.3 | XP_005162697.1 | 3 - 4 | 274 | 71 | ||
| XM_017353823.2 | XP_017209312.1 | 3 - 4 | 346 | 71 | ||
| XM_017353824.2 | XP_017209313.1 | 3 - 4 | 342 | 71 | ||
| GenBank mRNA | BC081421.1 | - | 4 - 5 | 391 | 71 | |
| BC164917.1 | - | 1 - 2 | 192 | 71 | ||
More Information
Set Membership: |
|

eloca
Genomic Map