Assay Details
| Gene Symbol: h2ax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005967.1 | NP_001005967.1 | 1 | 249 | 119 | |
| GenBank mRNA | BC083299.1 | - | 1 | 249 | 119 | |
| BC164227.1 | - | 1 | 211 | 119 | ||
More Information
Set Membership: |
|

h2ax1
Genomic Map