Assay Details
| Gene Symbol: efemp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008587.1 | NP_001008587.1 | 7 - 8 | 952 | 71 | |
| GenBank mRNA | BC086840.1 | - | 7 - 8 | 952 | 71 | |
More Information
Set Membership: |
|

efemp2a
Genomic Map